SY3H3D6759B8 Synthonix Corporation

3-Aminothieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD03783349
  • Cas Number : 58327-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47894.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47894.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart