SY3H3D6759BC Synthonix Corporation

2-Amino-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD01569537
  • Cas Number : 83265-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48000.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48000.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart