SY3H3D6759C4 Synthonix Corporation

4-Amino-2-fluoro-5-methoxybenzonitrile

  • MDL number MFCD26407017
  • Cas Number : 1441723-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48375.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48375.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart