SY3H3D6759F4 Synthonix Corporation

2-Amino-2-(4-chlorophenyl)acetic acid

  • MDL number MFCD00049328
  • Cas Number : 6212-33-5
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48955.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48955.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart