SY3H3D6759FF Synthonix Corporation

1-(2-Amino-4-bromophenyl)ethanone

  • MDL number MFCD16659002
  • Cas Number : 123858-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystal
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49024.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49024.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart