SY3H3D675A17 Synthonix Corporation

2-Amino-4-methylbenzoic acid

  • MDL number MFCD00047853
  • Cas Number : 2305-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49282.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49282.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart