SY3H3D675A18 Synthonix Corporation

3-Amino-5-methylbenzoic acid

  • MDL number No MDL found
  • Cas Number : 2305-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49283.html

Safety Information

MSDS https://synthonix.com/49283.html
Availability details for
Quantity
  • Availability
  • Add to Cart