SY3H3D675A1A Synthonix Corporation

3-Amino-4-methoxybenzaldehyde

  • MDL number MFCD07787578
  • Cas Number : 351003-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49285.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49285.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart