SY3H3D675A23 Synthonix Corporation

2-Amino-2-(3,4-dichlorophenyl)ethanol

  • MDL number MFCD12827189
  • Cas Number : 188586-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49366.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49366.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart