SY3H3D675A3D Synthonix Corporation

(S)-2-Amino-2-cyclohexylacetic acid hydrochloride

  • MDL number MFCD00050539
  • Cas Number : 191611-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49569.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49569.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart