SY3H3D675A43 Synthonix Corporation

3-Aminophthalic acid hydrochloride dihydrate

  • MDL number MFCD04971193
  • Cas Number : 1852533-96-0
  • Molecular Weight : 253.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49608.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49608.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart