SY3H3D675A5A Synthonix Corporation

(6R,7R)-7-Amino-8-oxo-3-vinyl-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid

  • MDL number MFCD07782149
  • Cas Number : 79349-82-9
  • Molecular Weight : 226.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49741.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49741.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart