SY3H3D675A65 Synthonix Corporation

2-Amino-3-(2-chlorophenyl)propanoic acid

  • MDL number MFCD00037774
  • Cas Number : 14091-11-3
  • Molecular Weight : 199.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49818.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49818.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart