SY3H3D675A69 Synthonix Corporation

3-Amino-3-(2-fluorophenyl)propanoic acid

  • MDL number MFCD00193241
  • Cas Number : 117391-49-8
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49834.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49834.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart