SY3H3D675A6D Synthonix Corporation

(R)-2-Amino-3-(4-fluorophenyl)propanoic acid

  • MDL number MFCD00063063
  • Cas Number : 18125-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49839.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49839.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart