SY3H3D67689C Synthonix Corporation

2-Amino-2-(3-(trifluoromethyl)phenyl)acetic acid hydrochloride

  • MDL number MFCD07371458
  • Cas Number : 1134915-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52225.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52225.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart