SY3H3D67689D Synthonix Corporation

2-(4-Amino-3-chlorophenyl)butanoic acid

  • MDL number MFCD24487840
  • Cas Number : 25814-13-5
  • Molecular Weight : 213.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52760.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52760.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart