SY3H3D67689E Synthonix Corporation

[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate cyclohexanamine

  • MDL number MFCD00058338
  • Cas Number : 102029-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form crystalline
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52912.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52912.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart