SY3H3D6768A0 Synthonix Corporation

3-Amino-1-phenylpropan-1-ol oxalate

  • MDL number MFCD29905488
  • Cas Number : 114843-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55426.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55426.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart