SY3H3D6768A2 Synthonix Corporation

2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid dihydrochloride

  • MDL number MFCD19440796
  • Cas Number : 132210-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56639.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56639.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart