SY3H3D6768A3 Synthonix Corporation

3-Amino-3-(4-fluoronaphthalen-1-yl)propanoic acid hydrochloride

  • MDL number MFCD28986163
  • Cas Number : 1810070-00-8
  • Molecular Weight : 269.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57404.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57404.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart