SY3H3D6768BA Synthonix Corporation

4-Acetamidobenzylamine

  • MDL number No MDL found
  • Cas Number : 25412-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50169.html

Safety Information

MSDS https://synthonix.com/50169.html
Availability details for
Quantity
  • Availability
  • Add to Cart