SY3H3D6768BB Synthonix Corporation

4-Amino-N,N-dimethylbenzamide

  • MDL number MFCD00523648
  • Cas Number : 6331-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50174.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50174.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart