SY3H3D6768C1 Synthonix Corporation

2-Amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD00111277
  • Cas Number : 4815-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50204.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50204.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart