SY3H3D6768C6 Synthonix Corporation

2-Amino-3-(2-chlorophenyl)propan-1-ol

  • MDL number MFCD10034618
  • Cas Number : 37844-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50258.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50258.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart