SY3H3D67694C Synthonix Corporation

2-Amino-2-(3-(trifluoromethyl)phenyl)acetic acid

  • MDL number MFCD02662404
  • Cas Number : 242475-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51477.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51477.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart