SY3H3D67695B Synthonix Corporation

3-Amino-3-(4-bromo-3-fluorophenyl)propanoic acid hydrochloride

  • MDL number MFCD30609570
  • Cas Number : 2061980-53-6
  • Molecular Weight : 298.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51793.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51793.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart