SY3H3D676962 Synthonix Corporation

(S)-2-Amino-3-(3,4-difluorophenyl)propanoic acid

  • MDL number MFCD01075125
  • Cas Number : 31105-90-5
  • Molecular Weight : 201.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/51812.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/51812.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart