SY3H3D6769DB Synthonix Corporation

tert-butyl 3-aminocyclobutane-1-carboxylate hydrochloride

  • MDL number MFCD21604762
  • Cas Number : 1192549-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52208.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52208.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart