SY3H3D6769E3 Synthonix Corporation

2-Amino-2-(1H-indol-5-yl)acetic acid

  • MDL number MFCD06656882
  • Cas Number : 108763-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Light-red or Yellow to Brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52309.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52309.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart