SY3H3D676A0D Synthonix Corporation

Adamantane-2,6-dione

  • MDL number MFCD01321093
  • Cas Number : 39751-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52786.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52786.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart