SY3H3D676A6E Synthonix Corporation

2-Amino-3-(2-methoxyphenyl)propanoic acid

  • MDL number MFCD03412504
  • Cas Number : 22976-68-7
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52981.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52981.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart