SY3H3D676A72 Synthonix Corporation

(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid

  • MDL number MFCD00070368
  • Cas Number : 59554-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52987.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52987.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart