SY3H3D676A73 Synthonix Corporation

4-Amino-5-(ethylthio)-2-methoxybenzoic acid

  • MDL number MFCD04973620
  • Cas Number : 71675-86-0
  • Molecular Weight : 227.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52991.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52991.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart