SY3H3D676A74 Synthonix Corporation

4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid

  • MDL number MFCD04973619
  • Cas Number : 71675-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/52998.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/52998.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart