SY3H3D676A82 Synthonix Corporation

2,2'-((4-Aminophenyl)azanediyl)diethanol sulfate

  • MDL number MFCD00013177
  • Cas Number : 54381-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53197.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53197.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart