SY3H3D676A8F Synthonix Corporation

2-Aminodecanoic acid

  • MDL number MFCD01862887
  • Cas Number : 17702-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53348.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53348.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart