SY3H3D676A91 Synthonix Corporation

5-Amino-1-(2-bromophenyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD03407394
  • Cas Number : 71856-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53550.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53550.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart