SY3H3D676AA0 Synthonix Corporation

2-Acetoxy-2',4'-difluoroacetophenone

  • MDL number MFCD09743655
  • Cas Number : 122263-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53814.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53814.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart