SY3H3D676AB3 Synthonix Corporation

(R)-3-Amino-4-(3,4-difluorophenyl)butanoic acid hydrochloride

  • MDL number MFCD01860951
  • Cas Number : 332061-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54114.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54114.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart