SY3H3D676B4E Synthonix Corporation

3-Amino-3-(3-methoxyphenyl)propanenitrile hydrochloride

  • MDL number MFCD30180244
  • Cas Number : 1956324-43-8
  • Molecular Weight : 212.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54119.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54119.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart