SY3H3D676B60 Synthonix Corporation

2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide

  • MDL number MFCD10000630
  • Cas Number : 302964-24-5
  • Molecular Weight : 267.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54304.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54304.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart