SY3H3D676B64 Synthonix Corporation

2-Amino-3-(5-chloro-1H-indol-3-yl)propanoic acid

  • MDL number MFCD01074499
  • Cas Number : 154-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54384.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54384.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart