SY3H3D676B7A Synthonix Corporation

1-(6-Amino-5-methoxyindolin-1-yl)ethanone

  • MDL number MFCD17392806
  • Cas Number : 23772-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54704.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54704.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart