SY3H3D676BB9 Synthonix Corporation

(S)-4-Amino-2-(((benzyloxy)carbonyl)amino)butanoic acid

  • MDL number MFCD00077904
  • Cas Number : 62234-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56040.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/56040.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart