SY3H3D676BBD Synthonix Corporation

3-Amino-3-(4-isopropylphenyl)propionic acid

  • MDL number MFCD02663184
  • Cas Number : 117391-53-4
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56151.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56151.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart