SY3H3D676BC0 Synthonix Corporation

1-Acetyladamantane

  • MDL number MFCD00074739
  • Cas Number : 1660-04-4
  • Molecular Weight : 178.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/56215.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/56215.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart