SY3H3D676BCA Synthonix Corporation

N1-(4-Aminophenyl)benzene-1,4-diamine sulfate

  • MDL number MFCD00013113
  • Cas Number : 53760-27-3
  • Molecular Weight : 297.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Greyish blue - Purple
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56558.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56558.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart