SY3H3D676BD6 Synthonix Corporation

2-Amino-5-fluoro-N-phenylbenzamide

  • MDL number MFCD11101521
  • Cas Number : 60041-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Off-white to light red powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56700.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56700.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart