SY3H3D676BDC Synthonix Corporation

1-Allylnaphthalene

  • MDL number MFCD00021581
  • Cas Number : 2489-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56741.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56741.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart